Alfa Aesar™ 1,2-Dichloro-4-fluorobenzene, 98+%

Manufacturer: Alfa Aesar™ A1135718

Quantity 50g

 View more versions of this product

Catalog No. AAA1135718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1435-49-0
    Molecular Formula C6H3Cl2F
    Formula Weight 165
    MDL Number MFCD00018119
    Beilstein 1861278
    Synonym 3,4-Dichloro-1-fluorobenzene
    Chemical Name or Material 1,2-Dichloro-4-fluorobenzene

    Specifications

    Formula Weight 165
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H3Cl2F
    Beilstein 1861278
    Melting Point −1°C
    Quantity 50g
    MDL Number MFCD00018119
    Boiling Point 168° to 171°C
    Synonym 3,4-Dichloro-1-fluorobenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    CAS 1435-49-0
    Flash Point 64°C (147°F)
    Chemical Name or Material 1,2-Dichloro-4-fluorobenzene
    Assay ≥98%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit