Alfa Aesar™ 1,2-Dichlorobenzene-d4, 99% (Isotopic)

Manufacturer: Alfa Aesar™ 4228003

Quantity 1g

 View more versions of this product

Catalog No. AA4228003

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2199-69-1
    Formula Weight 151.03
    MDL Number MFCD00037106
    UN Number UN1591
    Chemical Name or Material 1,2-Dichlorobenzene-d4, Isotopic

    Specifications

    Formula Weight 151.03
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    UN Number UN1591
    Density 1.34g/mL
    Boiling Point 181°C
    Quantity 1g
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: O-DICHLOROBENZENE
    Melting Point −17°C
    Flash Point 65°C (150°F)
    Refractive Index 1.5509
    MDL Number MFCD00037106
    CAS 2199-69-1
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Chemical Name or Material 1,2-Dichlorobenzene-d4, Isotopic
    Assay 99%

    GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit