Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, 99.8%, for HPLC, Thermo Scientific Chemicals

Catalog Number p-4229166
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
ClCCCl
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
ClCCCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY
Melting Point -35.0°C
Absorbance 0.005 max. at 260nm, 0.005 max. at 270nm, 0.01 max. at 300nm, 0.01 max. at 400nm, 0.05 max. at 250nm, 0.15 max. at 240nm, 1 max. at 230nm
Color White
Acidity 0.0001 meq/g max.
Alkalinity 0.0001 meq/g max.
Density 1.2500g/mL
Boiling Point 81.0°C to 85.0°C
Quantity 1 L
Flash Point 13°C
Packaging Glass Bottle
Refractive Index 1.4440 to 1.4460
Assay Percent Range 99.75% min. (GC)
Residue after Evaporation 2ppm max.
Specific Gravity 1.25
Linear Formula ClCH2CH2Cl
Viscosity 0.8 mPa.s (20°C)
Water 0.02% Max.
Merck Index 15,3851
Solubility Information Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 98.96
Percent Purity 99.80%
Grade HPLC
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wa
EINECSNumber 203-458-1

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit