Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, ACS, 99+%, Alfa Aesar

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, H2O 0.03%

Manufacturer:  Alfa Aesar 39121K7

Quantity 4L

 View more versions of this product

Catalog No. AA39121K7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

107-06-2
>99%
98.96
EDC
ACS Grade
Liquid
ClCH2CH2Cl
MFCD00000963
1, 2-Dichloroethane

Specifications

Clear colorless
107-06-2
440°C (824°F)
>99%
98.96
EDC
1, 2-Dichloroethane
4L
Colorless liquid
Liquid
ClCH2CH2Cl
MFCD00000963
1, 2-Dichloroethane
ACS Grade

Safety and Handling

SDS & Safety Documents

H225-H350-H302-H315-H319-H335: Highly flammable liquid and vapour. May cause cancer. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-458-1

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit