Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, Anhydrous, Approx. 99.8%, Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
107-06-2
C2H4Cl2
98.95
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
ClCCCl
107-06-2
C2H4Cl2
98.95
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
ClCCCl

D1386, 107-06-2, C2H4Cl2

Spectrum™ 1,2-Dichloroethane, Anhydrous is among Spectrum Chemicals′s high quality inventory of research chemicals. Also known as Ethylene Dichloride or Ethylene dichloride, 1,2-Dichloroethane is a chemical compound and chlorinated hydrocarbon. It is a colorless liquid with a chloroform-like scent and sweet taste, and is most often used in the production of vinyl chloride. Anhydrous solvents are used when performing certain reactions where the presence of water can prevent a chemical reaction from happening or form undesirable products. Spectrum anhydrous chemical solvents are packaged in convenient sizes featuring a double closure, which includes an easy-slip-on vinyl cap with Teflon liner and screw-on phenolic cap. These items are packaged in specially designed septum-sealed bottles under nitrogen to maintain low water content and ensure product integrity.
CAS Max % 100%
UN Number UN1184
Percent Purity Approx. 99.8%
Packaging Amber Glass Bottle
Residue after Evaporation 0.03%
Quantity 100 mL