Alfa Aesar™ 1,2-Dichloroethane, Spectrophotometric Grade, 99+%

Manufacturer: Alfa Aesar™ 32462M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA32462M1

 N/A
    N/A

This unit of measure is invalid. 


    Chemical Identifiers

    CAS 107-06-2
    Formula Weight 98.96
    MDL Number MFCD00000963
    UN Number UN1184
    Beilstein 605264
    Merck Index 14,3797
    Synonym EDC; Ethylene chloride
    Chemical Name or Material 1,2-Dichloroethane
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 98.96
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point −35°C
    Beilstein 605264
    Merck Index 14,3797
    Health Hazard 1 Danger
    RTECS Number KI0525000
    Refractive Index 1.4448
    Flash Point 15° (59°F)
    Boiling Point 83°C
    Physical Form Liquid
    UN Number UN1184
    Synonym EDC; Ethylene chloride
    Density 1.256g/mL
    TSCA TSCA
    Quantity 4 x 1L
    MDL Number MFCD00000963
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE DICHLORIDE
    CAS 107-06-2
    Health Hazard 2 GHS H Statement
    H225-H350-H302-H315-H319-H335
    Highly flammable liquid and vapor.
    May cause cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 1,2-Dichloroethane
    Assay ≥99%
    Grade Spectrophotometric grade

    GHS H Statement
    H225-H350-H302-H315-H319-H335
    Highly flammable liquid and vapor.
    May cause cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit