Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, Spectrophotometric Grade, ACS, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
107-06-2
107-06-2
CAS Max % 1
Percent Purity 0.99
Grade Spectrophotometric
Packaging Amber Glass Bottle
Residue after Ignition 2e-05
Titratable Acid 0.0003 meq/g
Water 0.0003
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.