Alfa Aesar™ 1,2-Dichlorooctafluorocyclohexene, 97%

Manufacturer: Alfa Aesar™ B2263303

Quantity 1g

 View more versions of this product

Catalog No. AAB2263303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 336-19-6
    Molecular Formula C6Cl2F8
    Formula Weight 294.95
    MDL Number MFCD00039466
    UN Number UN2810
    Chemical Name or Material 1,2-Dichlorooctafluorocyclohexene

    Specifications

    Formula Weight 294.95
    MDL Number MFCD00039466
    UN Number UN2810
    Boiling Point 112° to 114°C
    Health Hazard 2 GHS H Statement
    H330
    Fatal if inhaled.
    Health Hazard 3 GHS P Statement
    P280f-P304+P340-P309-P310
    Wear protective clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    RTECS Number GW6000000
    Quantity 1g
    Molecular Formula C6Cl2F8
    CAS 336-19-6
    Health Hazard 1 Danger
    Chemical Name or Material 1,2-Dichlorooctafluorocyclohexene
    Assay 97%

    GHS H Statement
    H330
    Fatal if inhaled.

    GHS P Statement
    P280f-P304+P340-P309-P310
    Wear protective clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit