Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dicyanobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic bottle
91-15-6
C8H4N2
128.13
MFCD00001771
XQZYPMVTSDWCCE-UHFFFAOYSA-N
phthalonitrile,1,2-dicyanobenzene,o-phthalodinitrile,phthalodinitrile,1,2-benzenedicarbonitrile,o-dicyanobenzene,1,2-benzodinitrile,o-pdn,o-cyanobenzonitrile,o-benzenedicarbonitrile
7042
benzene-1,2-dicarbonitrile
C1=CC=C(C(=C1)C#N)C#N
91-15-6
C8H4N2
128.13
MFCD00001771
XQZYPMVTSDWCCE-UHFFFAOYSA-N
phthalonitrile,1,2-dicyanobenzene,o-phthalodinitrile,phthalodinitrile,1,2-benzenedicarbonitrile,o-dicyanobenzene,1,2-benzodinitrile,o-pdn,o-cyanobenzonitrile,o-benzenedicarbonitrile
7042
benzene-1,2-dicarbonitrile
C1=CC=C(C(=C1)C#N)C#N
Specific Gravity 1.24
Packaging Glass bottle
Density 1.2400g/mL
Infrared Spectrum Authentic
Quantity 5kg
Color White to Beige
Flash Point >130°C
Melting Point 138.0°C to 142.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H4(CN)2
Beilstein 09, 815
Solubility Information Solubility in water: 0.56g/L (25°C). Other solubilities: soluble in acetone and benzene,soluble in ether and alcohol
Formula Weight 128.13
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1, 2-Dicyanobenzene, 98%
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger

EINECSNumber : 202-044-8

RTECSNumber : TI8575000

TSCA : TSCA