Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diethoxybenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2050-46-6
C10H14O2
166.22
MFCD00015143
QZYDOKBVZJLQCK-UHFFFAOYSA-N
o-diethoxybenzene,benzene, 1,2-diethoxy,catechol diethyl ether,benzene, o-diethoxy,o-diethoxy benzene,benzene, diethoxy,unii-p9dm5t78pz,pyrocatechol diethyl ether,p9dm5t78pz,diethoxybenzene-1,2
74904
1,2-diethoxybenzene
CCOC1=CC=CC=C1OCC
2050-46-6
C10H14O2
166.22
MFCD00015143
QZYDOKBVZJLQCK-UHFFFAOYSA-N
o-diethoxybenzene,benzene, 1,2-diethoxy,catechol diethyl ether,benzene, o-diethoxy,o-diethoxy benzene,benzene, diethoxy,unii-p9dm5t78pz,pyrocatechol diethyl ether,p9dm5t78pz,diethoxybenzene-1,2
74904
1,2-diethoxybenzene
CCOC1=CC=CC=C1OCC
Melting Point 40°C to 44°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H4(OC2H5)2
Formula Weight 166.22
Physical Form Crystalline Low Melting Mass or Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1, 2-Diethoxybenzene, 98%
Show More Show Less

EINECSNumber : 218-089-1

RTECSNumber : CZ5580000

TSCA : TSCA