Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Difluorobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
367-11-3
C6H4F2
114.09
MFCD00000284
GOYDNIKZWGIXJT-UHFFFAOYSA-N
o-difluorobenzene,benzene, 1,2-difluoro,benzene, o-difluoro,ortho-difluorobenzene,o-difluoro benzene,difluorobenzene,1,2-difluoro-benzene,unii-aw7qgmw29c,aw7qgmw29c,0-difluorobenzene
9706
CHEBI:38583
1,2-difluorobenzene
C1=CC=C(C(=C1)F)F
367-11-3
C6H4F2
114.09
MFCD00000284
GOYDNIKZWGIXJT-UHFFFAOYSA-N
o-difluorobenzene,benzene, 1,2-difluoro,benzene, o-difluoro,ortho-difluorobenzene,o-difluoro benzene,difluorobenzene,1,2-difluoro-benzene,unii-aw7qgmw29c,aw7qgmw29c,0-difluorobenzene
9706
CHEBI:38583
1,2-difluorobenzene
C1=CC=C(C(=C1)F)F
Packaging Glass bottle
Flash Point 2°C
Specific Gravity 1.158
Melting Point -34°C
Boiling Point 92°C
Density 1.1580g/mL
Infrared Spectrum Authentic
Refractive Index 1.4417 to 1.4437
Quantity 50mL
Color Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H4F2
Beilstein 05, IV, 637
Formula Weight 114.09
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1, 2-Difluorobenzene, 98%
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-680-7

RTECSNumber : CZ5655000

TSCA : TSCA