Learn More
1,2-Diiodobenzene, 98%, Thermo Scientific Chemicals
Description
1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.
Solubility
Slightly miscible with water.
Notes
Light sensitive and air sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatibile with strong oxidizing agents, strong acids and bases.
Specifications
Density | 2.524 |
Boiling Point | 282°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.7165 |
Quantity | 25 g |
Beilstein | 1855315 |
Sensitivity | Light sensitive |
Solubility Information | Slightly miscible with water. |
Formula Weight | 329.91 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H315-H318-H335 |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
EINECSNumber | 210-425-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.