Alfa Aesar™ 1,2-Diiodobenzene, 98%

Manufacturer: Alfa Aesar™ B2442506

Quantity 5g

 View more versions of this product

Catalog No. AAB2442506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 615-42-9
    Molecular Formula C6H4I2
    Formula Weight 329.91
    MDL Number MFCD00001038
    Beilstein 1855315
    Synonym o-Diiodobenzene
    Chemical Name or Material 1,2-Diiodobenzene

    Specifications

    Formula Weight 329.91
    Quantity 5g
    MDL Number MFCD00001038
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    TSCA TSCA
    Boiling Point 282°C
    Molecular Formula C6H4I2
    Flash Point >110°C (230°F)
    CAS 615-42-9
    Beilstein 1855315
    Synonym o-Diiodobenzene
    Health Hazard 1 Warning
    Chemical Name or Material 1,2-Diiodobenzene
    Assay 98%

    Description

    Applications
    1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.

    Solubility
    Slightly miscible with water.

    Notes
    Light sensitive and air sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatibile with strong oxidizing agents, strong acids and bases.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit