Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dipiperidinoethane, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1932-04-3
C12H24N2
196.33
MFCD00014644
UYMQPNRUQXPLCY-UHFFFAOYSA-N
1,2-dipiperidinoethane,1,2-dipiperidinylethane,piperidine, 1,1'-1,2-ethanediyl bis,1-2-piperidin-1-ylethyl piperidine,1,2-di piperidin-1-yl ethane,1,2-bis piperidino ethane,piperidine, 1,1'-ethylenedi,1-2-piperidin-1-yl ethyl piperidine,1,1'-ethylenedipiperidine,piperidine,1'-ethylenedi
74733
1932-04-3
C12H24N2
196.33
MFCD00014644
UYMQPNRUQXPLCY-UHFFFAOYSA-N
1,2-dipiperidinoethane,1,2-dipiperidinylethane,piperidine, 1,1'-1,2-ethanediyl bis,1-2-piperidin-1-ylethyl piperidine,1,2-di piperidin-1-yl ethane,1,2-bis piperidino ethane,piperidine, 1,1'-ethylenedi,1-2-piperidin-1-yl ethyl piperidine,1,1'-ethylenedipiperidine,piperidine,1'-ethylenedi
74733
Boiling Point 265°C
Melting Point -1°C to 0.5°C
Flash Point 110°C
Specific Gravity 0.91
Density 0.9100g/mL
Refractive Index 1.4865 to 1.4885
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Undesignated
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 20, I, 19
Formula Weight 196.33
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1, 2-Dipiperidinoethane, 98%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a posi

GHS Signal Word: Warning

EINECSNumber : 217-692-7

TSCA : TSCA