Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxy-5-hexene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
10353-53-4
C6H10O
98.15
MFCD00010051
MUUOUUYKIVSIAR-UHFFFAOYSA-N
1,2-epoxy-5-hexene,diallyl monoxide,2-3-butenyl oxirane,diallyl monooxide,oxirane, 3-butenyl,3-butenyloxirane,1-hexene, 5,6-epoxy,2-but-3-en-1-yl oxirane,5,6-epoxyhex-1-ene,ccris 3748
66314
2-but-3-enyloxirane
C=CCCC1CO1
10353-53-4
C6H10O
98.15
MFCD00010051
MUUOUUYKIVSIAR-UHFFFAOYSA-N
1,2-epoxy-5-hexene,diallyl monoxide,2-3-butenyl oxirane,diallyl monooxide,oxirane, 3-butenyl,3-butenyloxirane,1-hexene, 5,6-epoxy,2-but-3-en-1-yl oxirane,5,6-epoxyhex-1-ene,ccris 3748
66314
2-but-3-enyloxirane
C=CCCC1CO1
Specific Gravity 0.87
Boiling Point 118°C to 121°C
Flash Point 15°C
Refractive Index 1.4240 to 1.426
Quantity 25g
Density 0.8700g/mL
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: .. Other solubilities: soluble in aliphatic solvents,halogenated,solvents and alcohols
Formula Weight 98.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1, 2-Epoxy-5-hexene
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Danger

EINECSNumber : 233-771-9