Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxy-7-octene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
19600-63-6
C8H14O
126.199
MFCD00005156
UKTHULMXFLCNAV-UHFFFAOYSA-N
1,2-epoxy-7-octene,oxirane, 5-hexenyl,5-hexenyloxirane,7,8-epoxyoctene,ccris 3749,2-hex-5-en-1-yl oxirane,oxirane, 5-hexenyl-, s,2-hex-5-en-yloxirane,2-5-hexenyl oxirane,acmc-20ap64
29678
2-hex-5-enyloxirane
C=CCCCCC1CO1
19600-63-6
C8H14O
126.199
MFCD00005156
UKTHULMXFLCNAV-UHFFFAOYSA-N
1,2-epoxy-7-octene,oxirane, 5-hexenyl,5-hexenyloxirane,7,8-epoxyoctene,ccris 3749,2-hex-5-en-1-yl oxirane,oxirane, 5-hexenyl-, s,2-hex-5-en-yloxirane,2-5-hexenyl oxirane,acmc-20ap64
29678
2-hex-5-enyloxirane
C=CCCCCC1CO1
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4345 to 1.4365
Quantity 25g
Specific Gravity 0.85
Color Colorless
Flash Point 38°C
Density 0.8500g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: immiscible.
Formula Weight 126.2
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1, 2-Epoxy-7-octene, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-178-7

RTECSNumber : RR0502000

TSCA : TSCA