Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxyhexane, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
1436-34-6
C6H12O
100.16
MFCD00005154
WHNBDXQTMPYBAT-UHFFFAOYSA-N
1,2-epoxyhexane,butyloxirane,oxirane, butyl,1-hexene oxide,1-hexene epoxide,1,2-hexene oxide,epoxy-n-hexane,hexylene oxide,hexane, 1,2-epoxy,hexene, monooxide
15036
2-butyloxirane
CCCCC1CO1
1436-34-6
C6H12O
100.16
MFCD00005154
WHNBDXQTMPYBAT-UHFFFAOYSA-N
1,2-epoxyhexane,butyloxirane,oxirane, butyl,1-hexene oxide,1-hexene epoxide,1,2-hexene oxide,epoxy-n-hexane,hexylene oxide,hexane, 1,2-epoxy,hexene, monooxide
15036
2-butyloxirane
CCCCC1CO1
Refractive Index 1.4040 to 1.407
Specific Gravity 0.83
Boiling Point 118°C to 120°C
Flash Point 15°C
Color Colorless
Infrared Spectrum Authentic
Quantity 25mL
Packaging Glass bottle
Density 0.8300g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 100.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1, 2-Epoxyhexane
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 215-864-6