Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Ethanedisulfonic Acid, Spectrum™

Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle

E3520, 5982-56-9, C2H6O6S2

Spectrum™ 1,2-Ethanedisulfonic Acid , also known as Ethane-1,2-disulfonic acid, is a diprotic sulfonic acid used to form salts with the active ingredient in pharmaceutical formulations to increase aqueous solubility. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 25 g
CAS 5982-56-9
CAS Max % 1
Molecular Formula C2H10O8S2
UN Number UN2585
InChI Key DSGUSEBCDAKBCM-UHFFFAOYSA-N
SMILES O.O.OS(=O)(=O)CCS(O)(=O)=O
IUPAC Name ethane-1,2-disulfonic acid dihydrate
Molecular Weight (g/mol) 226.21