Alfa Aesar™ 1-(2-Ethoxyethyl)piperazine, 97%

Manufacturer: Alfa Aesar™ H55186MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55186MD

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 13484-38-3
    Formula Weight 158.24
    MDL Number MFCD01320899
    Chemical Name or Material 1-(2-Ethoxyethyl)piperazine

    Specifications

    Formula Weight 158.24
    Boiling Point 224° to 225°C
    Refractive Index 1.4690
    MDL Number MFCD01320899
    CAS 13484-38-3
    Color Colorless to dark yellow
    Quantity 250mg
    Physical Form Liquid
    Density 0.941g/mL
    Chemical Name or Material 1-(2-Ethoxyethyl)piperazine
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit