Alfa Aesar™ 1-(2-Ethoxyethyl)piperazine, 97%

Manufacturer: Alfa Aesar™ H5518603

Quantity 1g

 View more versions of this product

Catalog No. AAH5518603

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13484-38-3
    Formula Weight 158.24
    MDL Number MFCD01320899
    Chemical Name or Material 1-(2-Ethoxyethyl)piperazine

    Specifications

    Formula Weight 158.24
    Boiling Point 224° to 225°C
    Physical Form Liquid
    Refractive Index 1.4690
    CAS 13484-38-3
    Color Colorless to dark yellow
    Quantity 1g
    MDL Number MFCD01320899
    Density 0.941g/mL
    Chemical Name or Material 1-(2-Ethoxyethyl)piperazine
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit