Alfa Aesar™ 1-(2-Fluorobenzyl)piperazine, 96%

Manufacturer: Alfa Aesar™ H5588803

Quantity 1g

 View more versions of this product

Catalog No. AAH5588803

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 89292-78-4
    Formula Weight 194.25
    MDL Number MFCD02256027
    Chemical Name or Material 1-(2-Fluorobenzyl)piperazine

    Specifications

    Formula Weight 194.25
    Boiling Point 93° to 96°C (0.2mmHg)
    MDL Number MFCD02256027
    CAS 89292-78-4
    Physical Form Liquid
    Refractive Index 1.5330
    Density 1.170g/mL
    Color Colorless to pale brown
    Quantity 1g
    Chemical Name or Material 1-(2-Fluorobenzyl)piperazine
    Assay 96%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit