Alfa Aesar™ 1-(2-Fluorophenyl)ethanol, 95%

Manufacturer: Alfa Aesar™ H3190018

Quantity 50g

 View more versions of this product

Catalog No. AAH3190018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 445-26-1
    Molecular Formula C8H9FO
    Formula Weight 140.16
    MDL Number MFCD00014401
    Chemical Name or Material 1-(2-Fluorophenyl)ethanol

    Specifications

    Formula Weight 140.16
    Molecular Formula C8H9FO
    MDL Number MFCD00014401
    CAS 445-26-1
    Quantity 50g
    Chemical Name or Material 1-(2-Fluorophenyl)ethanol
    Assay 95%

    Description

    Applications
    It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

    Solubility
    Immiscible in water.

    Notes
    Store in cool dry place. Keep away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit