Alfa Aesar™ 1-(2-Fluorophenyl)propanol, 95%

Manufacturer: Alfa Aesar™ H3192918

Quantity 50g

 View more versions of this product

Catalog No. AAH3192918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 156022-15-0
    Molecular Formula C9H11FO
    Formula Weight 154.18
    MDL Number MFCD08062368
    Chemical Name or Material 1-(2-Fluorophenyl)propanol

    Specifications

    Formula Weight 154.18
    Molecular Formula C9H11FO
    MDL Number MFCD08062368
    Quantity 50g
    CAS 156022-15-0
    Chemical Name or Material 1-(2-Fluorophenyl)propanol
    Assay 95%

    Description

    Applications
    It is used as an intermediate in organic synthesis and is a pharmaceutical intermediate.

    Notes
    Store in cool dry place. Keep away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit