Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Hydroxyethyl)piperidine, 99%

Click to view available options
Quantity:
100g
500g
3040-44-6
C7H15NO
129.203
MFCD00006512
KZTWONRVIPPDKH-UHFFFAOYSA-N
2-piperidinoethanol,1-piperidineethanol,2-piperidin-1-yl ethanol,1-2-hydroxyethyl piperidine,n-2-hydroxyethyl piperidine,n-piperidineethanol,2-1-piperidinyl ethanol,n-hydroxyethyl piperidine,beta-piperidylethanol,beta-piperidinoethanol
18232
CHEBI:61238
2-piperidin-1-ylethanol
C1CCN(CC1)CCO
3040-44-6
C7H15NO
129.203
MFCD00006512
KZTWONRVIPPDKH-UHFFFAOYSA-N
2-piperidinoethanol,1-piperidineethanol,2-piperidin-1-yl ethanol,1-2-hydroxyethyl piperidine,n-2-hydroxyethyl piperidine,n-piperidineethanol,2-1-piperidinyl ethanol,n-hydroxyethyl piperidine,beta-piperidylethanol,beta-piperidinoethanol
18232
CHEBI:61238
2-piperidin-1-ylethanol
C1CCN(CC1)CCO
Melting Point 16°C
Odor Amine-like
Quantity 100g
Boiling Point 199°C to 202°C
pH 12.4
Flash Point 83°C (181°F)
UN Number UN2735
Beilstein 103390
Formula Weight 129.2
Percent Purity 99%
Chemical Name or Material 1-(2-Hydroxyethyl)piperidine
Show More Show Less

GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : TM8052000

TSCA : TSCA