Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Methoxyethyl)piperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13484-40-7
C7H16N2O
144.21
MFCD00191214
BMEMBBFDTYHTLH-UHFFFAOYSA-N
1-2-methoxyethyl piperazine,1-2-methoxyethyl-piperazine,1-2-methoxy-ethyl-piperazine,piperazine, 1-2-methoxyethyl,1-methoxy-2-piperazinylethane,pubchem14209,2-methoxyethylpiperazine,n-methoxyethyl piperazine,2-methoxyethyl piperazine,n-methoxyethylylpiperazine
2734638
1-(2-methoxyethyl)piperazine
COCCN1CCNCC1
13484-40-7
C7H16N2O
144.21
MFCD00191214
BMEMBBFDTYHTLH-UHFFFAOYSA-N
1-2-methoxyethyl piperazine,1-2-methoxyethyl-piperazine,1-2-methoxy-ethyl-piperazine,piperazine, 1-2-methoxyethyl,1-methoxy-2-piperazinylethane,pubchem14209,2-methoxyethylpiperazine,n-methoxyethyl piperazine,2-methoxyethyl piperazine,n-methoxyethylylpiperazine
2734638
1-(2-methoxyethyl)piperazine
COCCN1CCNCC1
Refractive Index 1.4730 to 1.475
Density 0.9700g/mL
Boiling Point 193°C to 194°C
Flash Point 87°C
Specific Gravity 0.97
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 144.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-(2-Methoxyethyl)piperazine, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger