Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Methoxyphenyl)piperazine 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
35386-24-4
C11H16N2O
192.26
MFCD00005958
VNZLQLYBRIOLFZ-UHFFFAOYSA-N
1-2-methoxyphenyl piperazine,1-2-methoxy-phenyl-piperazine,1-o-methoxyphenyl piperazine,2-mpp,1-2-methoxyphenyl-piperazine,n-2-methoxyphenyl piperazine,unii-81njo1330a,1-2-methoxy phenyl piperazine,2-methoxyphenylpiperazine,piperazine, 1-2-methoxyphenyl
1346
1-(2-methoxyphenyl)piperazine
COC1=CC=CC=C1N2CCNCC2
35386-24-4
C11H16N2O
192.26
MFCD00005958
VNZLQLYBRIOLFZ-UHFFFAOYSA-N
1-2-methoxyphenyl piperazine,1-2-methoxy-phenyl-piperazine,1-o-methoxyphenyl piperazine,2-mpp,1-2-methoxyphenyl-piperazine,n-2-methoxyphenyl piperazine,unii-81njo1330a,1-2-methoxy phenyl piperazine,2-methoxyphenylpiperazine,piperazine, 1-2-methoxyphenyl
1346
1-(2-methoxyphenyl)piperazine
COC1=CC=CC=C1N2CCNCC2
Melting Point 35.0°C to 38.0°C
Density 1.0900g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5745 to 1.5765
Quantity 25g
Specific Gravity 1.09
Color Colorless to Yellow
Boiling Point 130.0°C to 133.0°C (0.1mmHg)
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Solubility Information Solubility in water: insoluble
Formula Weight 192.26
Physical Form Liquid After Melting
Percent Purity ≥98%
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine, 98+%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 252-537-7

TSCA : TSCA