Alfa Aesar™ 1,2-Methylenedioxy-4-nitrobenzene, 98+%

Manufacturer: Alfa Aesar™ B2483814

Quantity 25g

 View more versions of this product

Catalog No. AAB2483814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2620-44-2
    Molecular Formula C7H5NO4
    Formula Weight 167.12
    MDL Number MFCD00005824
    Beilstein 169063
    Synonym 5-Nitro-1,3-benzodioxole; 4-Nitro-1,2-(methylenedioxy)benzene
    Chemical Name or Material 1,2-Methylenedioxy-4-nitrobenzene

    Specifications

    Formula Weight 167.12
    TSCA TSCA
    MDL Number MFCD00005824
    CAS 2620-44-2
    Synonym 5-Nitro-1,3-benzodioxole; 4-Nitro-1,2-(methylenedioxy)benzene
    Melting Point 142° to 149°C
    Quantity 25g
    Flash Point 142°C (287°F)
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H5NO4
    Beilstein 169063
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Chemical Name or Material 1,2-Methylenedioxy-4-nitrobenzene
    Assay ≥98%

    GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit