Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[2-(Morpholin-4-yl)ethyl]piperazine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4892-89-1
C10H21N3O
199.29
MFCD00191217
SAJZEJMFAWZNCQ-UHFFFAOYSA-N
4-2-piperazin-1-yl-ethyl-morpholine,4-2-piperazin-1-yl ethyl morpholine,4-2-piperazin-1-ylethyl morpholine,1-2-morpholin-4-yl-ethyl-piperazine,1-2-morpholinoethyl-piperazine,1-2-morpholin-4-yl ethyl piperazine,morpholine, 4-2-1-piperazinyl ethyl,1-2-morpholinoethyl piperazine,4-2-1-piperazinyl ethyl morpholine
2734640
4-(2-piperazin-1-ylethyl)morpholine
C1CN(CCN1)CCN2CCOCC2
4892-89-1
C10H21N3O
199.29
MFCD00191217
SAJZEJMFAWZNCQ-UHFFFAOYSA-N
4-2-piperazin-1-yl-ethyl-morpholine,4-2-piperazin-1-yl ethyl morpholine,4-2-piperazin-1-ylethyl morpholine,1-2-morpholin-4-yl-ethyl-piperazine,1-2-morpholinoethyl-piperazine,1-2-morpholin-4-yl ethyl piperazine,morpholine, 4-2-1-piperazinyl ethyl,1-2-morpholinoethyl piperazine,4-2-1-piperazinyl ethyl morpholine
2734640
4-(2-piperazin-1-ylethyl)morpholine
C1CN(CCN1)CCN2CCOCC2
Boiling Point 82°C to 84°C (0.5 mbar)
Infrared Spectrum Authentic
Packaging Glass bottle
Color White to Yellow
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 199.29
Physical Form Low Melting Crystals or Oil
Percent Purity 99%
Chemical Name or Material 1-[2-(Morpholin-4-yl)ethyl]piperazine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger