Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-N-BOC-aminoethyl)piperazine, 95%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
140447-78-5
C11H23N3O2
229.324
VPOIPCJBJNWHSJ-UHFFFAOYSA-N
1-2-n-boc-aminoethyl piperazine,1-2-boc-aminoethyl piperazine,1-2-boc-amino ethyl piperazine,tert-butyl 2-piperazin-1-yl ethyl carbamate,tert-butyl 2-piperazin-1-yl ethylcarbamate,2-piperazin-1-yl-ethyl-carbamic acid tert-butyl ester,tert-butyl n-2-piperazin-1-yl ethyl carbamate,tert-butyl n-2-piperazin-1-ylethyl carbamate,carbamic acid, n-2-1-piperazinyl ethyl-, 1,1-dimethylethyl ester,carbamic acid, 2-1-piperazinyl ethyl-, 1,1-dimethylethyl ester
1514401
tert-butyl N-(2-piperazin-1-ylethyl)carbamate
CC(C)(C)OC(=O)NCCN1CCNCC1
140447-78-5
C11H23N3O2
229.324
VPOIPCJBJNWHSJ-UHFFFAOYSA-N
1-2-n-boc-aminoethyl piperazine,1-2-boc-aminoethyl piperazine,1-2-boc-amino ethyl piperazine,tert-butyl 2-piperazin-1-yl ethyl carbamate,tert-butyl 2-piperazin-1-yl ethylcarbamate,2-piperazin-1-yl-ethyl-carbamic acid tert-butyl ester,tert-butyl n-2-piperazin-1-yl ethyl carbamate,tert-butyl n-2-piperazin-1-ylethyl carbamate,carbamic acid, n-2-1-piperazinyl ethyl-, 1,1-dimethylethyl ester,carbamic acid, 2-1-piperazinyl ethyl-, 1,1-dimethylethyl ester
1514401
tert-butyl N-(2-piperazin-1-ylethyl)carbamate
CC(C)(C)OC(=O)NCCN1CCNCC1
Quantity 250mg
Infrared Spectrum Authentic
Formula Weight 229.32
Percent Purity 95%
Chemical Name or Material 1-(2-N-BOC-aminoethyl)piperazine

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

Warning