Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Naphthoquinone, 95%, Tech., ACROS Organics™

Click to view available options
Quantity:
1g
5g
10g
25g
Packaging:
Glass bottle
524-42-5
C10H6O2
158.16
MFCD00001698
KETQAJRQOHHATG-UHFFFAOYSA-N
1,2-naphthoquinone,1,2-naphthalenedione,o-naphthoquinone,beta-naphthoquinone,1,2-naphthaquinone,1,2-naftochinon,.beta.-naphthoquinone,1,2-naftochinon czech,ccris 1558,1,2-dihydronaphthalene-1,2-dione
10667
CHEBI:34055
naphthalene-1,2-dione
C1=CC=C2C(=C1)C=CC(=O)C2=O
524-42-5
C10H6O2
158.16
MFCD00001698
KETQAJRQOHHATG-UHFFFAOYSA-N
1,2-naphthoquinone,1,2-naphthalenedione,o-naphthoquinone,beta-naphthoquinone,1,2-naphthaquinone,1,2-naftochinon,.beta.-naphthoquinone,1,2-naftochinon czech,ccris 1558,1,2-dihydronaphthalene-1,2-dione
10667
CHEBI:34055
naphthalene-1,2-dione
C1=CC=C2C(=C1)C=CC(=O)C2=O
Melting Point 136.0°C to 141.0°C
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color Colorless or White to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Beilstein 07, 709
Fieser 01,713
Merck Index 15, 6479
Solubility Information Solubility in water: negligible. Other solubilities: soluble in 5% naoh,5% nahco3 and conc. h2so4,soluble in alcohol,benzene and ether
Formula Weight 158.16
Physical Form Crystalline Mass or Waxy Solid
Percent Purity 95%
Grade Technical
Chemical Name or Material 1, 2-Naphthoquinone, tech., 97%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 208-360-2

RTECSNumber : QL7000000

TSCA : TSCA