Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Nitrophenyl)piperidine, 98%

Click to view available options
Quantity:
5g
25g
100g
15822-77-2
C11H14N2O2
206.245
MFCD00023653
WDAUAKBDZSYXEM-UHFFFAOYSA-N
1-2-nitrophenyl piperidine,2-nitrophenyl piperidine,pubchem10243,maybridge1_000639,acmc-1c4am,cbmicro_018648,cambridge id 5345369,1-o-nitrophenyl piperidine,1-2-nitrophenyl piperidine #,piperidine,1-2-nitrophenyl
225457
1-(2-nitrophenyl)piperidine
C1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
15822-77-2
C11H14N2O2
206.245
MFCD00023653
WDAUAKBDZSYXEM-UHFFFAOYSA-N
1-2-nitrophenyl piperidine,2-nitrophenyl piperidine,pubchem10243,maybridge1_000639,acmc-1c4am,cbmicro_018648,cambridge id 5345369,1-o-nitrophenyl piperidine,1-2-nitrophenyl piperidine #,piperidine,1-2-nitrophenyl
225457
1-(2-nitrophenyl)piperidine
C1CCN(CC1)C2=CC=CC=C2[N+](=O)[O-]
Melting Point 76°C to 78°C
Quantity 100g
Formula Weight 206.24
Percent Purity 98%
Chemical Name or Material 1-(2-Nitrophenyl)piperidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning