Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Pentanediol, 96%, Acros Organics™

Click to view available options
Quantity:
50mL
250mL
Packaging:
Glass bottle
5343-92-0
C5H12O2
104.15
WCVRQHFDJLLWFE-UHFFFAOYSA-N
1,2-pentanediol,1,2-dihydroxypentane,1,2-pentanediol, 2r,acmc-20mbh5,acmc-1axdb,3-01-00-02191 beilstein handbook reference,ksc271i3n,wcvrqhfdjllwfe-uhfffaoysa
93000
pentane-1,2-diol
CCCC(CO)O
5343-92-0
C5H12O2
104.15
WCVRQHFDJLLWFE-UHFFFAOYSA-N
1,2-pentanediol,1,2-dihydroxypentane,1,2-pentanediol, 2r,acmc-20mbh5,acmc-1axdb,3-01-00-02191 beilstein handbook reference,ksc271i3n,wcvrqhfdjllwfe-uhfffaoysa
93000
pentane-1,2-diol
CCCC(CO)O
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250mL
Specific Gravity 0.971
Refractive Index 1.4387 to 1.4407
Color Colorless to Yellow
Flash Point 105°C
Density 0.9710g/mL
Boiling Point 206.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula CH3(CH2)2CH(OH)CH2OH
Beilstein 01,II,548
Solubility Information Solubility in water: miscible.
Formula Weight 104.15
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 1, 2-Pentanediol, 97%
Show More Show Less

GHS P Statement
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 226-285-3

RTECSNumber : SA0455000

TSCA : TSCA