Alfa Aesar™ 1,2-Phenylene phosphorochloridite, 97%

Manufacturer: Alfa Aesar™ B2271214

Quantity 25g

 View more versions of this product

Catalog No. AAB2271214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1641-40-3
    Molecular Formula C6H4ClO2P
    Formula Weight 174.52
    MDL Number MFCD00005853
    UN Number UN3261
    Beilstein 135455
    Synonym Catechyl phosphorus chloride; 2-Chloro-1,3,2-benzodioxaphosphole
    Chemical Name or Material 1,2-Phenylene phosphorochloridite

    Specifications

    Formula Weight 174.52
    TSCA TSCA
    UN Number UN3261
    Beilstein 135455
    Synonym Catechyl phosphorus chloride; 2-Chloro-1,3,2-benzodioxaphosphole
    Health Hazard 1 Danger
    Melting Point ≈30°C
    Flash Point >110°C (230°F)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    Molecular Formula C6H4ClO2P
    MDL Number MFCD00005853
    CAS 1641-40-3
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 73° to 75°C (7mmHg)
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 1,2-Phenylene phosphorochloridite
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit