Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Phthalic Dicarboxaldehyde, +98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
643-79-8
C8H6O2
134.13
MFCD00003335
ZWLUXSQADUDCSB-UHFFFAOYSA-N
o-phthalaldehyde,o-phthaldialdehyde,benzene-1,2-dicarboxaldehyde,1,2-benzenedicarboxaldehyde,phthaldialdehyde,phthalic aldehyde,phthalic dialdehyde,phthalyldicarboxaldehyde,ortho-phthalaldehyde,o-phthaldehyde
4807
CHEBI:70851
phthalaldehyde
C1=CC=C(C(=C1)C=O)C=O
643-79-8
C8H6O2
134.13
MFCD00003335
ZWLUXSQADUDCSB-UHFFFAOYSA-N
o-phthalaldehyde,o-phthaldialdehyde,benzene-1,2-dicarboxaldehyde,1,2-benzenedicarboxaldehyde,phthaldialdehyde,phthalic aldehyde,phthalic dialdehyde,phthalyldicarboxaldehyde,ortho-phthalaldehyde,o-phthaldehyde
4807
CHEBI:70851
phthalaldehyde
C1=CC=C(C(=C1)C=O)C=O
Boiling Point 83°C to 84°C (1.0 hPa)
Viscosity 2.3 mPa.s (65°C)
Melting Point 54.0°C to 57.0°C
Color White
Packaging Glass bottle
Flash Point 132°C
Infrared Spectrum Authentic
Quantity 100g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula C6H4(CHO)2
Beilstein 07, 674
Merck Index 15, 7480
Solubility Information Solubility in water: soluble.
Formula Weight 134.13
Physical Form Powder
Percent Purity ≥98%
Chemical Name or Material 1, 2-Phthalic dicarboxaldehyde
Show More Show Less

GHS H Statement
Toxic if swallowed. Causes severe skin burns and eye damage. May cause an allergic skin reaction. Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid release to the environment.

Danger