Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Propanediol, ACS, 99.5%, Alfa Aesar

As a substitute for ethylene glycol and glycerol

Manufacturer:  Alfa Aesar 309481Q

Quantity 55 gal.

 View more versions of this product

Catalog No. AA309481Q

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

57-55-6
99.5%
76.1
ACS Grade
Liquid
CH3CH(OH)CH2OH
1, 2-Propanediol

Specifications

57-55-6
Liquid
CH3CH(OH)CH2OH
186 to 188°
107°C
1, 2-Propanediol
1.036g/mL
99.5%
76.1
−60°C (−76°F)
55 gal.
ACS Grade

Safety and Handling

storageNote1 : Hygroscopic

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit