1,2-Propanediol, ACS, 99.5%, Alfa Aesar

As a substitute for ethylene glycol and glycerol

Manufacturer: Alfa Aesar 30948AP

Quantity 500mL

 View more versions of this product

Catalog No. AA30948AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57-55-6
    Physical Form Liquid
    Assay Percent Range 99.5%
    Molecular Formula CH3CH(OH)CH2OH
    Formula Weight 76.1
    Chemical Name or Material 1, 2-Propanediol
    Grade ACS Grade

    Specifications

    Formula Weight 76.1
    Quantity 500mL
    Physical Form Liquid
    Density 1.036
    Assay Percent Range 99.5%
    CAS 57-55-6
    Storage Note 1 Hygroscopic
    Molecular Formula CH3CH(OH)CH2OH
    Chemical Name or Material 1, 2-Propanediol
    Grade ACS Grade

    Hygroscopic

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit