Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Propanediol, Reagent, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
57-55-6
C3H8O2
76.10
DNIAPMSPPWPWGF-UHFFFAOYNA-N
propane-1,2-diol
CC(O)CO
57-55-6
C3H8O2
76.10
DNIAPMSPPWPWGF-UHFFFAOYNA-N
propane-1,2-diol
CC(O)CO

P1440, 57-55-6, C3H8O2

Spectrum™ 1,2-Propanediol, Reagent, ACS or propylene glycol is an organic compound and colorless liquid without odor. It has a slightly sweet taste and chemically classed as a diol. It is considered generally recognized as safe (GRAS) by the U.S. Food and Drug Administration, and it is used as an humectant (E1520), solvent, and preservative in food and for tobacco products, as well as being one of the major ingredients of the "e-liquid" used in electronic cigarettes along with vegetable glycerin. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Water 0.2%
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 100%
Percent Purity 99.5%