Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Pyrimidinyl)piperazine dihydrochloride, 98+%

Click to view available options
Quantity:
5g
25g
100g
94021-22-4
C8H14Cl2N4
237.128
MFCD00012779
ZNZGJSLHXOMREP-UHFFFAOYSA-N
2-1-piperazinyl pyrimidine dihydrochloride,1-2-pyrimidyl piperazine dihydrochloride,2-piperazin-1-yl pyrimidine dihydrochloride,1-2-pyrimidinyl piperazine dihydrochloride,2-piperazinylpyrimidine hcl,2-piperazinylpyrimidine dihydrochloride,2-piperazin-1-ylpyrimidine dihydrochloride,pyrimidine, 2-1-piperazinyl-, dihydrochloride,pyrimidinylpiperazine dihydrochloride,pubchem7349
11680272
2-piperazin-1-ylpyrimidine;dihydrochloride
C1CN(CCN1)C2=NC=CC=N2.Cl.Cl
94021-22-4
C8H14Cl2N4
237.128
MFCD00012779
ZNZGJSLHXOMREP-UHFFFAOYSA-N
2-1-piperazinyl pyrimidine dihydrochloride,1-2-pyrimidyl piperazine dihydrochloride,2-piperazin-1-yl pyrimidine dihydrochloride,1-2-pyrimidinyl piperazine dihydrochloride,2-piperazinylpyrimidine hcl,2-piperazinylpyrimidine dihydrochloride,2-piperazin-1-ylpyrimidine dihydrochloride,pyrimidine, 2-1-piperazinyl-, dihydrochloride,pyrimidinylpiperazine dihydrochloride,pubchem7349
11680272
2-piperazin-1-ylpyrimidine;dihydrochloride
C1CN(CCN1)C2=NC=CC=N2.Cl.Cl
Melting Point ~280°C (decomposition)
Quantity 5g
Beilstein 151178
Formula Weight 237.13
Percent Purity ≥98%
Chemical Name or Material 1-(2-Pyrimidinyl)piperazine dihydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UV9702000