Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-(2-Pyrimidinyl)piperazine dihydrochloride, 98+%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Sensitivity | Hygroscopic |
Quantity | 25g |
Melting Point | ~280°C (decomposition) |
Beilstein | 151178 |
Formula Weight | 237.13 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-(2-Pyrimidinyl)piperazine dihydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 301-415-2
RTECSNumber : UV9702000
TSCA : No
Recommended Storage : Ambient temperatures