Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(2-Thienyl)piperidine, 98%

Click to view available options
Quantity:
250mg
1g
5g
19983-20-1
C9H13NS
167.27
MFCD00142961
SVUBKHLEMUNAQT-UHFFFAOYSA-N
1-thiophen-2-yl piperidine,1-thien-2-yl-piperidine,piperidine,1-2-thienyl,2-piperidylthiophene,thienylpiperidine,2-piperidinothiophene,n-2-thienyl piperidine,1-thien-2-yl piperidine,2-piperidino-thiophene,1-2-thienyl piperidine
12181160
1-thiophen-2-ylpiperidine
C1CCN(CC1)C2=CC=CS2
19983-20-1
C9H13NS
167.27
MFCD00142961
SVUBKHLEMUNAQT-UHFFFAOYSA-N
1-thiophen-2-yl piperidine,1-thien-2-yl-piperidine,piperidine,1-2-thienyl,2-piperidylthiophene,thienylpiperidine,2-piperidinothiophene,n-2-thienyl piperidine,1-thien-2-yl piperidine,2-piperidino-thiophene,1-2-thienyl piperidine
12181160
1-thiophen-2-ylpiperidine
C1CCN(CC1)C2=CC=CS2
Color Orange
Sensitivity Light Sensitive
Quantity 250mg
Beilstein 1239029
Formula Weight 167.27
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 1-(2-Thienyl)piperidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning