Alfa Aesar™ 1-[2-(Trifluoromethoxy)phenyl]ethanol, 97%

Manufacturer: Alfa Aesar™ H3224414

Quantity 25g

 View more versions of this product

Catalog No. AAH3224414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 910442-37-4
    Molecular Formula C9H9F3O2
    Formula Weight 206.16
    MDL Number MFCD08062367
    Chemical Name or Material 1-[2-(Trifluoromethoxy)phenyl]ethanol

    Specifications

    Formula Weight 206.16
    Quantity 25g
    Molecular Formula C9H9F3O2
    MDL Number MFCD08062367
    CAS 910442-37-4
    Chemical Name or Material 1-[2-(Trifluoromethoxy)phenyl]ethanol
    Assay 97%

    Description

    Applications
    Used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit