Learn More
Saf T Pak Inc STP-104 - Reusable Secondary Pressure Receptacle with Orange Lid
Supplier: Saf T Pak Inc STP104
STP-104 Reusable Secondary Pressure Receptacle with Orange Lid, 1.25L, Outer Demensions: 6.25 x 4.25in., Polypropylene Threaded Lid with O-Ring, 6/Case