Learn More
Estes Industries, LLC 1/2A3-4T Engines



The Estes 1/2A3-4T is a single stage model rocket engine with a short delay ejection charge for use in small, lightweight model rockets. This engine is 13mm in diameter and intended for launching model rockets weighing less than 2 ounces, including the engine. Each package includes 4 engines, 4 starters and 4 mini starter plugs.
Supplier: Estes Industries, LLC 001504
Description
- The Estes 1/2A3-4T is a single stage model rocket engine with a short delay ejection charge for use in small, lightweight model rockets.
- This engine is 13 mm in diameter and intended for launching model rockets weighing less than 2 ounces, including the engine.
- Each package includes 4 engines, 4 starters and 4 mini starter plugs.
Specifications
Brown | |
0.5 in. | |
Rocket | |
4 1/2A3-4 T Engines, 4 Starters, 4 Starter Plugs | |
4.6 cm | |
0.23 oz. | |
0.23 in. |
Model Rocket Engine | |
Rocket Engine | |
1.8 in. | |
1.8 in. | |
Cardboard | |
6.6 g |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.