Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1(2H)-Phthalazinone, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
119-39-1
C8H6N2O
146.15
MFCD00006892
IJAPPYDYQCXOEF-UHFFFAOYSA-N
1 2h-phthalazinone,phthalazin-1 2h-one,phthalazone,phthalazinone,phthalazin-1-one,1-phthalazinol,phthalazin-1-ol,1-hydroxyphthalazine,1 2h phthalazinone,1,2-dihydrophthalazin-1-one
8394
CHEBI:34023
2H-phthalazin-1-one
C1=CC=C2C(=C1)C=NNC2=O
119-39-1
C8H6N2O
146.15
MFCD00006892
IJAPPYDYQCXOEF-UHFFFAOYSA-N
1 2h-phthalazinone,phthalazin-1 2h-one,phthalazone,phthalazinone,phthalazin-1-one,1-phthalazinol,phthalazin-1-ol,1-hydroxyphthalazine,1 2h phthalazinone,1,2-dihydrophthalazin-1-one
8394
CHEBI:34023
2H-phthalazin-1-one
C1=CC=C2C(=C1)C=NNC2=O
Melting Point 184.0°C to 187.0°C
Color White to Yellow
Infrared Spectrum Authentic
Quantity 100g
Packaging Glass bottle
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (HPLC)
Formula Weight 146.15
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 1(2H)-Phthalazinone
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 204-319-8