Learn More
Medchemexpress LLC 1-[(2R,6S)-6-(hydroxymethyl)-4-tritylmorpholin-2-yl]-5-methylpyrimidine-2,4-dione | 914361-76-5 | MFCD22666300 | 95.0% | 483.56 g/mol | C29H29N3O4 | 1 G

Supplier: Medchemexpress LLC HYW3572021G
7'-OH-N-trityl morpholinothymine is a nucleoside analog used as a PMO (phosphorodiamidate morpholino oligomer) thymidine precursor for research and chemical synthesis. It is supplied as a research-grade small molecule with characterized identity and supporting documentation for laboratory use.
- CAS number: 914361-76-5
- Molecular formula: C29H29N3O4
- Molecular weight: 483.56 g/mol
- Purity: 95.0%
- Available in multiple pack sizes, including 1 G
- Intended for research and synthesis of nucleic acid analogs
- Datasheet and certificate of analysis available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.