Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,4-Dimethylphenyl)piperazine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1014-05-7
C12H18N2
190.29
SFLNVAVCCYTHCQ-UHFFFAOYSA-N
1-3,4-dimethylphenyl piperazine,1-3,4-dimethylphenyl-piperazine,1-3,4-xylyl piperazine,1-3,4-dimethyl-phenyl-piperazine,piperazine, 1-3,4-dimethylphenyl,3,4-dimethylphenyl piperazine,pubchem9390,acmc-20a4ek,1-3,4-dimethyl-phenyl piperazine,1-3,4-dimethyl phenyl piperazine
2723782
1-(3,4-dimethylphenyl)piperazine
CC1=C(C=C(C=C1)N2CCNCC2)C
1014-05-7
C12H18N2
190.29
SFLNVAVCCYTHCQ-UHFFFAOYSA-N
1-3,4-dimethylphenyl piperazine,1-3,4-dimethylphenyl-piperazine,1-3,4-xylyl piperazine,1-3,4-dimethyl-phenyl-piperazine,piperazine, 1-3,4-dimethylphenyl,3,4-dimethylphenyl piperazine,pubchem9390,acmc-20a4ek,1-3,4-dimethyl-phenyl piperazine,1-3,4-dimethyl phenyl piperazine
2723782
1-(3,4-dimethylphenyl)piperazine
CC1=C(C=C(C=C1)N2CCNCC2)C
Water 0.5% max.
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Beige to Yellow
Melting Point 60°C to 65°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 190.29
Physical Form Crystalline Powder
Percent Purity 98.0 to 102.0% Total base
Chemical Name or Material 1-(3, 4-Dimethylphenyl)piperazine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger