Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3,5,5-Tetramethyl-1,3-cyclohexadiene, 94%

Click to view available options
Quantity:
1g
5g
4724-89-4
C10H16
136.238
MFCD00045518
SZHAWDAGEJWQJK-UHFFFAOYSA-N
1,3,5,5-tetramethyl-1,3-cyclohexadiene,1,3-cyclohexadiene, 1,3,5,5-tetramethyl,acmc-1alu2,1, 1,3,5,5-tetramethyl,1,3,5,5-tetramethylcyclohexadiene,1,5,5-tetramethyl-1,3-cyclohexadiene,1,3-cyclohexadiene,1,3,5,5-tetramethyl
78453
1,3,5,5-tetramethylcyclohexa-1,3-diene
CC1=CC(=CC(C1)(C)C)C
4724-89-4
C10H16
136.238
MFCD00045518
SZHAWDAGEJWQJK-UHFFFAOYSA-N
1,3,5,5-tetramethyl-1,3-cyclohexadiene,1,3-cyclohexadiene, 1,3,5,5-tetramethyl,acmc-1alu2,1, 1,3,5,5-tetramethyl,1,3,5,5-tetramethylcyclohexadiene,1,5,5-tetramethyl-1,3-cyclohexadiene,1,3-cyclohexadiene,1,3,5,5-tetramethyl
78453
1,3,5,5-tetramethylcyclohexa-1,3-diene
CC1=CC(=CC(C1)(C)C)C
Quantity 5g
Flash Point 52°C (126°F)
UN Number UN3295
Formula Weight 136.24
Percent Purity 94%
Chemical Name or Material 1,3,5,5-Tetramethyl-1,3-cyclohexadiene

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : TSCA