Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,5-Anhydro-2-deoxy-beta-D-threo-pentofuranosyl)thymine 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
7481-90-5
C10H12N2O4
224.216
OAWLMYIJZBBZTP-XLPZGREQSA-N
3′C,5′C-Anhydrothymidine
14069470
1-[(1S,3R,5R)-4,7-dioxabicyclo[3.2.0]heptan-3-yl]-5-methylpyrimidine-2,4-dione
CC1=CN(C(=O)NC1=O)C2CC3C(O2)CO3
7481-90-5
C10H12N2O4
224.216
OAWLMYIJZBBZTP-XLPZGREQSA-N
3′C,5′C-Anhydrothymidine
14069470
1-[(1S,3R,5R)-4,7-dioxabicyclo[3.2.0]heptan-3-yl]-5-methylpyrimidine-2,4-dione
CC1=CN(C(=O)NC1=O)C2CC3C(O2)CO3
Color White
Quantity 5g
Melting Point 196°C
Formula Weight 224.22
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 1-(3,5-Anhydro-2-deoxy-beta-D-threo-pentofuranosyl)thymine

EINECSNumber : (5)-6691

TSCA : No