Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,3,5-Benzenetricarboxylic acid triallyl ester, 98%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
Specifications
Melting Point | 31°C to 34°C |
Boiling Point | 212°C (2.0mmHg) |
Flash Point | >113°C |
Specific Gravity | 1.16 |
Density | 1.1600g/mL |
Quantity | 5g |
Packaging | Glass bottle |
Color | Yellow |
Infrared Spectrum | Authentic |
CAS Min % | 97.5 |
Show More |
Safety and Handling
EINECSNumber : 241-791-4