Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(3,5-Dichlorophenyl)-2,5-dimethylpyrrole, 97%

Manufacturer:  Alfa Aesar™ B2512506

Quantity 5g

 View more versions of this product

Catalog No. AAB2512506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

175205-50-2
240.13
1-(3,5-Dichlorophenyl)-2,5-dimethylpyrrole
C12H11Cl2N
MFCD00084999

Specifications

5g
C12H11Cl2N
MFCD00084999
1-(3,5-Dichlorophenyl)-2,5-dimethylpyrrole
175205-50-2
240.13
78° to 80°C
97%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit